• Inicio
  • Resultados de la búsqueda
  • Genetic Effects of eNOS Polymorphisms on Biomarkers Related to Cardiovascular Status in a Population Coexposed to Methylmercury and Lead.
Genetic Effects of eNOS Polymorphisms on Biomarkers Related to Cardiovascular Status in a Population Coexposed to Methylmercury and Lead.
Archives of environmental contamination and toxicology (2015-02-19)
Gustavo Rafael Mazzaron Barcelos, Kátia Cristina De Marco, Vânia Braghini de Rezende, Gilberto Úbida Leite Braga, Lusânia Maria Greggi Antunes, José Eduardo Tanus-Santos, Fernando Barbosa
RESUMEN
The aim of the present study was to evaluate possible effects of endothelial nitric oxide synthase (eNOS) polymorphisms on systolic (SBP) and diastolic blood pressure (DBP) and on nitrite levels in plasma (NitP) in a population coexposed to methylhemoglobin (MeHg) and lead (Pb) in the Amazonian region, Brazil. Plasmatic levels of hemoglobin Hg (HgP) and Pb (PbP) were determined by inductively coupled plasma-mass spectrometry, whereas NitP were quantified by chemiluminescence. Genotyping was performed by conventional and restriction fragment length polymorphism-polymerase chain reaction assay. The population age ranged from 18 to 87 years (mean 40 ± 16), and the distribution between the sexes was homogenous (63 men and 50 women). Mean HgP and PbP were 7.1 ± 6.1 and 1.1 ± 1.1 µg L(-1), respectively. PbP was correlated to SBP and DBP, whereas no effects were observed for HgP on blood pressure. Subjects carrying the 4b allele in intron 4 presented greater SBP and DBP compared with those who had the 4a4a genotype. In addition, interactions between alcohol consumption and the -786 T/C polymorphism were observed on NitP, i.e., individuals carrying the polymorphic allele and drinkers had lower NitP. Taken together, our data give new insights concerning the genetic effects of eNOS polymorphisms on biomarkers related to cardiovascular status in populations coexposed to Hg and Pb.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%