Merck
  • Inicio
  • Resultados de la búsqueda
  • Versatile preparation of intracellular-acidity-sensitive oxime-linked polysaccharide-doxorubicin conjugate for malignancy therapeutic.
Versatile preparation of intracellular-acidity-sensitive oxime-linked polysaccharide-doxorubicin conjugate for malignancy therapeutic.
Biomaterials (2015-04-25)
Weiguo Xu, Jianxun Ding, Chunsheng Xiao, Lingyu Li, Xiuli Zhuang, Xuesi Chen
RESUMEN
Recently, chemotherapy has been one of the most important therapeutic approaches for malignant tumors. The tumor tissular or intracellular microenvironment-sensitive polymer-doxorubicin (DOX) conjugates demonstrate great potential for improved antitumor efficacy and reduced side effects. In this work, the acid-sensitive dextran-DOX conjugate (noted as Dex-O-DOX) was synthesized through the versatile efficient oximation reaction between the terminal aldehyde group of polysaccharide and the amino group in DOX in the buffer solution of sodium acetate/acetic acid. The insensitive one, i.e., Dex-b-DOX, was prepared similarly as Dex-O-DOX with a supplemented reduction reaction. The DOX release from Dex-O-DOX was pH-dependent and accelerated by the decreased pH. The efficient intracellular DOX release from Dex-O-DOX toward the human hepatoma HepG2 cells was further confirmed. Furthermore, Dex-O-DOX exhibited a closer antiproliferative activity to free DOX·HCl as the extension of time. More importantly, compared with Dex-b-DOX, Dex-O-DOX exhibited higher antitumor activity and lower toxicity, which were further confirmed by the systemic histological and immunohistochemical analyses. Hence, the facilely prepared smart polysaccharide-DOX conjugates, i.e., Dex-O-DOX, exhibited great potential in the clinical chemotherapy of malignancy.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
SAFC
Acetic acid, glacial
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%