Merck
A detailed investigation on electro-Fenton treatment of propachlor: Mineralization kinetic and degradation intermediates.
Chemosphere (2015-05-20)
Metin Gençten, Ali Özcan
RESUMEN
In this study, electrochemical removal of propachlor from water has been investigated by electro-Fenton process which provides continuous electrochemical production of hydroxyl radical which is a highly oxidizing agent. This radical can react with propachlor unselectively and can oxidize it into carbon dioxide and water. Effects of applied current, catalyst (Fe2(SO4)3) and supporting electrolyte (Na2SO4) concentrations on the degradation rate of propachlor have been examined and determined as 200 mA, 0.20 mM and 25 mM, respectively. The oxidation reaction showed a second-order reaction kinetic with an absolute rate constant value of (3.6±0.2)×10(9) M(-1) s(-1) which was determined by competition kinetic experiments. Total organic carbon analysis was employed to follow the mineralization of propachlor. The total mineralization was completed in a seven-hour electrolysis at 300 mA indicating that the electro-Fenton process is very effective in the mineralization of propachlor in water. Oxidation of propachlor with hydroxyl radical led to the formation of intermediate species. Some of these species were detected and quantified by chromatographic and spectroscopic methods such as HPLC, GC-MS and IC. A plausible mineralization pathway for the electrochemical removal of propachlor was proposed based on the identified by-products.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ammonium nitrate, 99.999% trace metals basis
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)