  • Search Results
  • Sensitive characterization of polyphenolic antioxidants in Polygonatum odoratum by selective solid phase extraction and high performance liquid chromatography-diode array detector-quadrupole time-of-flight tandem mass spectrometry.
Sensitive characterization of polyphenolic antioxidants in Polygonatum odoratum by selective solid phase extraction and high performance liquid chromatography-diode array detector-quadrupole time-of-flight tandem mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2015-05-06)
Xin Hu, Huading Zhao, Shuyun Shi, Hui Li, Xiaoling Zhou, Feipeng Jiao, Xinyu Jiang, Dongming Peng, Xiaoqin Chen
ABSTRACT
The complexity of natural products always leads to the co-elution of interfering compounds with bioactive compounds, which then has a detrimental effect on structural elucidation. Here, a new method, based on selective solid phase extraction combined with 1,1-diphenyl-2-picrylhydrazyl radical (DPPH) spiking and high performance liquid chromatography-diode array detector-quadrupole time-of-flight tandem mass spectrometry (HPLC-DAD-QTOF-MS/MS), is described for sensitive screening, selective extraction and identification of polyphenolic antioxidants in Polygonatum odoratum. First, 25 polyphenolic antioxidants (1-25) were screened by DPPH spiking with HPLC. Second, polydopamine coated Fe3O4 microspheres (Fe3O4@PDA) were prepared to selectively extract target antioxidants with extraction efficiency from 55% to 100% when the amount of Fe3O4@PDA, extraction time, desorption solvent and time were 10mg, 20 min, acetonitrile, and 5 min. Third, 25 antioxidants (10 cinnamides and 15 homoisoflavanones) were identified by HPLC-DAD-QTOF-MS/MS. Furthermore, the DPPH scavenging activities of purified compounds (IC50, 1.6-32.8 μg/mL) validated the method. Among the identified antioxidants, four of them (12, 13, 18 and 19) were new compounds, four of them (2, 4, 8 and 14) were first obtained from family Liliaceae, five of them (1, 3, 5, 7 and 9) were first reported in genus Polygonatum, while one compound (24) was first identified in this species. The results indicated that the proposed method was an efficient and sensitive approach to explore polyphenolic antioxidants from complex natural products.
Sigma-Aldrich
Furfural, ≥98%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Furfural, natural, ≥98%, FCC, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Furfural, ACS reagent, 99%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Furfural, 99%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C