• Inicio
  • Resultados de la búsqueda
  • A newly established culture method highlights regulatory roles of retinoic acid on morphogenesis and calcification of mammalian limb cartilage.
A newly established culture method highlights regulatory roles of retinoic acid on morphogenesis and calcification of mammalian limb cartilage.
BioTechniques (2014-01-01)
Eizo Masuda, Kota Shirai, Kenji Maekubo, Yohei Hirai
RESUMEN
During mammalian embryogenesis, sclerotome-derived chondrocytes in the limb bud are arranged into a complicated bone shape with specific areas undergoing hypertrophy and calcification, creating a region-specific mineralized pattern in the cartilage. To follow chondrogenesis progression in vitro, we isolated limb cartilage from mice on embryonic day 13 (E13) and cultured it at the air-liquid interface after microsurgical removal of the ectoderm/epidermis. Explants underwent proper morphogenesis, giving rise to complete templates for limb bones in vitro. We found that region-specific calcification patterns resembling limbs of prepartum mature embryos could be induced in explants using culture medium containing high concentrations of CaCl2 (Ca), ascorbic acid (AA), and β-glycerophosphoric acid (BGP). In this culture system, excess amounts of all-trans retinoic acid (RA) severely disrupted morphogenesis and calcification patterns in limb cartilage. These effects were more pronounced in forearms than in phalanges. Although dissociated, the nascent chondrocytes in culture did not give rise to cartilage units even though augmented calcification was induced in these cell aggregates in the presence of RA. Taken together, our newly established culture system revealed that RA independently regulates three-dimensional morphogenesis and calcification.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Glycerol phosphate disodium salt hydrate, isomeric mixture
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
β-Glycerophosphate disodium salt hydrate, ≤1.0 mol % L-α-isomer
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture