Specific labeling of climbing fibers shows early synaptic interactions with immature Purkinje cells in the prenatal cerebellum.
Developmental neurobiology (2014-12-23)
Yoshiaki Kita, Kazuto Tanaka, Fujio Murakami
RESUMEN
During development, growing axons must locate target cells to form synapses. This is not easy, since target cells are also growing and even actively migrating. In some brain regions, such axons have been reported to wait for the timing when target cells become mature, without invading their target region. However, in the cerebellum climbing fibers (CFs), major afferent axons, arrive near their target neurons, Purkinje cells, when the neurons are still actively migrating. We, therefore, examined whether synaptic contacts are established at such early stages. To specifically label CFs, we introduced by in utero electroporation a mixture of genes encoding for Ptf1a-enhancer-driven Cre recombinase and Cre-dependent fluorescent protein into the mouse hindbrain at embryonic day (E) 10.5 and observed them during development. The earliest stages at which labeled CFs were observed in the cerebellar primordium were E15.5-E16.5. These fibers were fasciculated in the dorsal region and entered the cerebellar primordium. Some fibers defasciculated and reached the caudal region. At E17.5 and E18.5, fasciculated fibers were also found in the mantle region, and some grew toward the surface of the primordium to penetrate a mass of Purkinje cells. Interestingly, as early as E16.5, labeled fibers were found to run in close apposition to Purkinje cell dendrites and to express a presynaptic marker. These observations suggest that CFs form synapses with Purkinje cells as soon as the fibers enter the cerebellum.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Dabco® 33-LV
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture