Development of a Novel Tetravalent Synthetic Peptide That Binds to Phosphatidic Acid.
PloS one (2015-07-07)
Rina Ogawa, Kohjiro Nagao, Kentaro Taniuchi, Masaki Tsuchiya, Utako Kato, Yuji Hara, Takehiko Inaba, Toshihide Kobayashi, Yoshihiro Sasaki, Kazunari Akiyoshi, Miho Watanabe-Takahashi, Kiyotaka Nishikawa, Masato Umeda
RESUMEN
We employed a multivalent peptide-library screening technique to identify a peptide motif that binds to phosphatidic acid (PA), but not to other phospholipids such as phosphatidylcholine (PC), phosphatidylethanolamine (PE), and phosphatidylserine (PS). A tetravalent peptide with the sequence motif of MARWHRHHH, designated as PAB-TP (phosphatidic acid-binding tetravalent peptide), was shown to bind as low as 1 mol% of PA in the bilayer membrane composed of PC and cholesterol. Kinetic analysis of the interaction between PAB-TP and the membranes containing 10 mol% of PA showed that PAB-TP associated with PA with a low dissociation constant of KD = 38 ± 5 nM. Coexistence of cholesterol or PE with PA in the membrane enhanced the PAB-TP binding to PA by increasing the ionization of the phosphomonoester head group as well as by changing the microenvironment of PA molecules in the membrane. Amino acid replacement analysis demonstrated that the tryptophan residue at position 4 of PAB-TP was involved in the interaction with PA. Furthermore, a series of amino acid substitutions at positions 5 to 9 of PAB-TP revealed the involvement of consecutive histidine and arginine residues in recognition of the phosphomonoester head group of PA. Our results demonstrate that the recognition of PA by PAB-TP is achieved by a combination of hydrophobic, electrostatic and hydrogen-bond interactions, and that the tetravalent structure of PAB-TP contributes to the high affinity binding to PA in the membrane. The novel PA-binding tetravalent peptide PAB-TP will provide insight into the molecular mechanism underlying the recognition of PA by PA-binding proteins that are involved in various cellular events.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
SAFC
Cholesterol, Plant-Derived
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
O-Phospho-L-serine
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
1,2-Dimyristoyl-sn-glycero-3-phosphocholine, ≥99%
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, ≥97%
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Tris(tert-butoxy)silanol, packaged for use in deposition systems