A diagnostic approach for identifying anti-neuronal antibodies in children with suspected autoimmune encephalitis.
Journal of neuroimmunology (2015-07-23)
Eri Nakahara, Hiroshi Sakuma, Junko Kimura-Kuroda, Toshiaki Shimizu, Akihisa Okumura, Masaharu Hayashi
RESUMEN
We assessed the validity of immunoblotting, immunohistochemistry (IHC), and immunocytochemistry (ICC) to detect anti-neuronal antibodies in an attempt to establish a diagnostic approach for pediatric autoimmune encephalitis. Both IHC and ICC had higher sensitivity than immunoblotting and could differentiate between antibodies directed towards intracellular and cell surface antigens. There was a significant correlation between the IHC and ICC results. When patients were divided into encephalitis and non-encephalitis groups, there was no difference in the positivity rate and staining pattern of IHC and ICC between them. In conclusion, IHC and ICC are useful methods to screen for anti-neuronal antibodies. A combination of IHC, ICC, and specific cell-based assays is expected to be an efficient approach for the diagnosis of autoantibody-mediated encephalitis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
3,3′-Diaminobenzidine, ≥97% (HPLC)
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
3,3′-Diaminobenzidine, 98%
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
DL-Cysteine hydrochloride, ≥95% (TLC), anhydrous
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C