Merck
Evaluation of methamphetamine-associated socioeconomic status and addictive behaviors, and their impact on oral health.
Addictive behaviors (2015-07-08)
Niklas Rommel, Nils H Rohleder, Stefan Wagenpfeil, Roland Haertel-Petri, Marco R Kesting
RESUMEN
Chronic methamphetamine abuse can lead to multiple health hazards. In particular, the substance is associated with devastating effects on oral health including symptoms such as rampant caries, gingiva inflammation, and xerostomia, whereby the term "Meth Mouth" occurs in the current literature. However, "Meth Mouth" pathology is primarily described on the basis of individual cases or has been evaluated without consideration of the mass of potential influencing factors. Therefore, we have conducted a systematic study to investigate the effects of accompanying factors and circumstances on oral health in cases of chronic methamphetamine abuse. In cooperation with two centers for addiction medicine, we assessed the data of 100 chronic methamphetamine users and 100 matched-pair controls between March 2012 and November 2013. We investigated their socioeconomic status, details of methamphetamine consumption behavior, collateral consumption of sugar beverages, nicotine alcohol, and other addictive substances including cannabis, opioids, other stimulants, and hallucinogens, and dental care. We found considerably greater unstable social circumstances, a high collateral consumption of substances with pathogenic potential for the stomatognathic system, and significantly poorer dental care in the methamphetamine-user group. Various factors have to be considered with regard to methamphetamine use and its influence on oral health. These factors can trigger potential damage by the drug methamphetamine possibly leading to the symptoms of "Meth Mouth", and should be considered in prevention and therapy strategies.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%