• Inicio
  • Resultados de la búsqueda
  • Simultaneous detection of endocrine disrupting chemicals including conjugates in municipal wastewater and sludge with enhanced sample pretreatment and UPLC-MS/MS.
Simultaneous detection of endocrine disrupting chemicals including conjugates in municipal wastewater and sludge with enhanced sample pretreatment and UPLC-MS/MS.
Environmental science. Processes & impacts (2015-07-15)
Bing Zhu, Weiwei Ben, Xiangjuan Yuan, Yu Zhang, Min Yang, Zhimin Qiang
RESUMEN
The co-existence of free and conjugated estrogens and the interference from complex matrices often lead to largely variable detected concentrations and sometimes even negative removal efficiencies of typical endocrine disrupting chemicals (EDCs) in wastewater treatment plants (WWTPs). In this study, a highly selective and sensitive method was developed for simultaneous extraction, elution, and detection of 12 EDCs (i.e., 4 free estrogens, 6 conjugated estrogens, and 2 phenolic compounds) in municipal wastewater and sludge. Sample pretreatment and ultra-performance liquid chromatography-tandem mass spectrometry detection were optimized to improve the detection selectivity and sensitivity. The results indicate that the additional purification process was highly effective in reducing the matrix interference, and the limits of quantification reached as low as 0.04-2.2 ng L(-1) in wastewater and 0.05-4.9 ng g(-1) in sludge for all target EDCs. The developed method was successfully applied to explore the behavior of target EDCs in a local WWTP. The conjugates occupied a considerable portion (4.3-76.9% in molar ratio) of each related estrogen in the influent. Most of the target EDCs could not be completely removed in WWTPs, thus posing a potential threat to aquatic ecosystems.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Estrone, ≥99%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Estriol, ≥97%
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Estriol, meets USP testing specifications
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ammonium hydroxide solution, ≥25% NH3 in H2O, semiconductor grade VLSI PURANAL (Honeywell 17605)