Merck
  • Inicio
  • Resultados de la búsqueda
  • Determination of Nicotine in Tobacco by Chemometric Optimization and Cation-Selective Exhaustive Injection in Combination with Sweeping-Micellar Electrokinetic Chromatography.
Determination of Nicotine in Tobacco by Chemometric Optimization and Cation-Selective Exhaustive Injection in Combination with Sweeping-Micellar Electrokinetic Chromatography.
Journal of analytical methods in chemistry (2015-06-24)
Yi-Hui Lin, Chia-Hsien Feng, Shih-Wei Wang, Po-Yun Ko, Ming-Hsun Lee, Yen-Ling Chen
RESUMEN
Nicotine is a potent chemical that excites the central nervous system and refreshes people. It is also physically addictive and causes dependence. To reduce the harm of tobacco products for smokers, a law was introduced that requires tobacco product containers to be marked with the amount of nicotine as well as tar. In this paper, an online stacking capillary electrophoresis (CE) method with cation-selective exhaustive injection sweeping-micellar electrokinetic chromatography (CSEI-sweeping-MEKC) is proposed for the optimized analysis of nicotine in tobacco. A higher conductivity buffer (160 mM phosphate buffer (pH 3)) zone was injected into the capillary, allowing for the analytes to be electrokinetically injected at a voltage of 15 kV for 15 min. Using 50 mM sodium dodecyl sulfate and 25% methanol in the sweeping buffer, nicotine was detected with high sensitivity. Thus, optimized conditions adapted from a chemometric approach provided a 6000-fold increase in the nicotine detection sensitivity using the CSEI-sweeping-MEKC method in comparison to normal CZE. The limits of detection were 0.5 nM for nicotine. The stacking method in combination with direct injection which matrix components would not interfere with assay performance was successfully applied to the detection of nicotine in tobacco samples.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, 99.998% trace metals basis