MiR-155 is upregulated in patients with active tuberculosis and inhibits apoptosis of monocytes by targeting FOXO3.
Molecular medicine reports (2015-09-02)
Jian Huang, Junhua Jiao, Weihua Xu, Huayang Zhao, Chunxiao Zhang, Yan Shi, Zhijian Xiao
RESUMEN
The aim of the present study was to investigate the association between microRNA (miR)-155 and apoptosis of monocytes infected by Mycobacterium tuberculosis, to examine the effect of forkhead box O3 (FOXO3) on miR‑155. The present study analysed the apoptosis of CD14+ in the peripheral blood of patients with active tuberculosis, disposed the THP‑1 human monocytic cell line by BCG and examined the expression of miR‑155. Furthermore, the expression of FOXO3 in THP‑1 cells was determined, and wild- and mutant-type luciferase reporter plasmids containing FOXO3 3'‑untranslated regions (UTRs) were constructed to analyse the expression of luciferase. Finally, an over‑expression plasmid was constructed, and THP-1 cells were transfected with control miRNA, miR‑155 and the plasmid, which revealed that miR‑155 inhibited the apoptosis of THP‑1 cells. miR‑155 in the THP‑1 cells infected by BCG was upregulated and apoptosis also increased. However, the apoptosis declined when the cells were transfected with the control miRNA and miR‑155. Folllowing transfection with miR‑155, the expression of FOXO3 decreased. Transfection with miR‑155 and the FOXO3 3'-UTRs significantly reduced luciferase, and overexpression of FOXO3 reversed the inhibitory role of miR‑155. From these results, it was concluded that mycobacteria can improve the level of miR‑155, while BCG can induce apoptosis in THP‑1 cells. The results suggested FOXO3 is a downstream target gene of miR‑155, which combines 3'-UTRs to inhibit the expression of FOXO3.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Fluorescein (free acid), Dye content 95 %
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Palmitic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)