The molecular basis for development of proinflammatory autoantibodies to progranulin.
Journal of autoimmunity (2015-05-26)
Lorenz Thurner, Natalie Fadle, Evi Regitz, Maria Kemele, Philipp Klemm, Marina Zaks, Elisabeth Stöger, Birgit Bette, Gabi Carbon, Vincent Zimmer, Gunter Assmann, Niels Murawski, Boris Kubuschok, Gerhard Held, Klaus-Dieter Preuss, Michael Pfreundschuh
RESUMEN
Recently we identified in a wide spectrum of autoimmune diseases frequently occurring proinflammatory autoantibodies directed against progranulin, a direct inhibitor of TNFR1 & 2 and of DR3. In the present study we investigated the mechanisms for the breakdown of self-tolerance against progranulin. Isoelectric focusing identified a second, differentially electrically charged progranulin isoform exclusively present in progranulin-antibody-positive patients. Alkaline phosphatase treatment revealed this additional progranulin isoform to be hyperphosphorylated. Subsequently Ser81, which is located within the epitope region of progranulin-antibodies, was identified as hyperphosphorylated serine residue by site directed mutagenesis of candidate phosphorylation sites. Hyperphosphorylated progranulin was detected exclusively in progranulin-antibody-positive patients during the courses of their diseases. The occurrence of hyperphosphorylated progranulin preceded seroconversions of progranulin-antibodies, indicating adaptive immune response. Utilizing panels of kinase and phosphatase inhibitors, PKCβ1 was identified as the relevant kinase and PP1 as the relevant phosphatase for phosphorylation and dephosphorylation of Ser81. In contrast to normal progranulin, hyperphosphorylated progranulin interacted exclusively with inactivated (pThr320) PP1, suggesting inactivated PP1 to cause the detectable occurrence of phosphorylated Ser81 PGRN. Investigation of possible functional alterations of PGRN due to Ser81 phosphorylation revealed, that hyperphosphorylation prevents the interaction and thus direct inhibition of TNFR1, TNFR2 and DR3, representing an additional direct proinflammatory effect. Finally phosphorylation of Ser81 PGRN alters the conversion pattern of PGRN. In conclusion, inactivated PP1 induces hyperphosphorylation of progranulin in a wide spectrum of autoimmune diseases. This hyperphosphorylation prevents direct inhibition of TNFR1, TNFR2 and DR3 by PGRN, alters the conversion of PGRN, and is strongly associated with the occurrence of neutralizing, proinflammatory PGRN-antibodies, indicating immunogenicity of this alternative secondary modification.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl