Singly and binary grafted poly(vinyl chloride) urinary catheters that elute ciprofloxacin and prevent bacteria adhesion.
International journal of pharmaceutics (2015-04-22)
Luisa Islas, Carmen Alvarez-Lorenzo, Beatriz Magariños, Angel Concheiro, Luis Felipe del Castillo, Guillermina Burillo
RESUMEN
Acrylic acid (AAc) and poly(ethylene glycol) methacrylate (PEGMA) were singly and dually grafted onto poly(vinyl chloride) (PVC) urinary catheters with the aim of preventing biofouling by endowing the catheters with the ability to load and release antimicrobial agents and to avoid bacteria adhesion. The polymers were grafted applying an oxidative pre-irradiation ((60)Co source) method in two steps. Grafting percentage and kinetics were evaluated by varying the absorbed pre-irradiation dose, reaction time, monomer concentration, and reaction temperature. Catheters with grafting percentages ranging from 8 to 207% were characterized regarding thermal stability, surface hydrophilicity, mechanical properties, swelling, and lubricity. The modified catheters proved to have better compatibility with fibroblast cells than PVC after long exposure times. Furthermore, grafted catheters were able to load ciprofloxacin and sustained its release in urine medium for several hours. Ciprofloxacin-loaded catheters inhibited the growth of Escherichia coli and Staphylococcus aureus in the catheter surroundings and prevented bacteria adhesion.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium chloride, for molecular biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis