Merck
Genomics of Clostridium botulinum group III strains.
Research in microbiology (2014-08-12)
Yoshihiko Sakaguchi, Tomonori Suzuki, Yumiko Yamamoto, Atsushi Nishikawa, Keiji Oguma
ABSTRACT
In Clostridium botulinum, the characteristics of type C and D strains are quite different from other types, and they are classified as group III. They produce C2 binary toxin and C3 exoenzyme in addition to type C and D neurotoxins. Two different phages and many plasmids are identified in the organisms. The genes of neurotoxin and C3 exoenzyme are converted from toxigenic strains to non-toxigenic strains by the specific bacteriophages (phages), whereas, the C2 toxin gene is carried by large or small plasmids. Classification of type C and D strains has been in confusion because 1) antigenicity of type C and D neurotoxins is complex, 2) the cells produce two types of toxins, neurotoxin and C2 toxin, and 3) some non-toxigenic strains can be converted to produce C or D neurotoxin by the infection with phages. Until now, entire nucleotide sequences of cell chromosomes, phages, and plasmids have been determined. Since both genetic and protein-chemical analyses have been clarifying the above confusions, these data are reviewed historically.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%