Merck
  • Inicio
  • Resultados de la búsqueda
  • Bioactive magnetic near Infra-Red fluorescent core-shell iron oxide/human serum albumin nanoparticles for controlled release of growth factors for augmentation of human mesenchymal stem cell growth and differentiation.
Bioactive magnetic near Infra-Red fluorescent core-shell iron oxide/human serum albumin nanoparticles for controlled release of growth factors for augmentation of human mesenchymal stem cell growth and differentiation.
Journal of nanobiotechnology (2015-05-08)
Itay Levy, Ifat Sher, Enav Corem-Salkmon, Ofra Ziv-Polat, Amilia Meir, Avraham J Treves, Arnon Nagler, Ofra Kalter-Leibovici, Shlomo Margel, Ygal Rotenstreich
RESUMEN
Iron oxide (IO) nanoparticles (NPs) of sizes less than 50 nm are considered to be non-toxic, biodegradable and superparamagnetic. We have previously described the generation of IO NPs coated with Human Serum Albumin (HSA). HSA coating onto the IO NPs enables conjugation of the IO/HSA NPs to various biomolecules including proteins. Here we describe the preparation and characterization of narrow size distribution core-shell NIR fluorescent IO/HSA magnetic NPs conjugated covalently to Fibroblast Growth Factor 2 (FGF2) for biomedical applications. We examined the biological activity of the conjugated FGF2 on human bone marrow mesenchymal stem cells (hBM-MSCs). These multipotent cells can differentiate into bone, cartilage, hepatic, endothelial and neuronal cells and are being studied in clinical trials for treatment of various diseases. FGF2 enhances the proliferation of hBM-MSCs and promotes their differentiation toward neuronal, adipogenic and osteogenic lineages in vitro. The NPs were characterized by transmission electron microscopy, dynamic light scattering, ultraviolet-visible spectroscopy and fluorescence spectroscopy. Covalent conjugation of the FGF2 to the IO/HSA NPs significantly stabilized this growth factor against various enzymes and inhibitors existing in serum and in tissue cultures. IO/HSA NPs conjugated to FGF2 were internalized into hBM-MSCs via endocytosis as confirmed by flow cytometry analysis and Prussian Blue staining. Conjugated FGF2 enhanced the proliferation and clonal expansion capacity of hBM-MSCs, as well as their adipogenic and osteogenic differentiation to a higher extent compared with the free growth factor. Free and conjugated FGF2 promoted the expression of neuronal marker Microtubule-Associated Protein 2 (MAP2) to a similar extent, but conjugated FGF2 was more effective than free FGF2 in promoting the expression of astrocyte marker Glial Fibrillary Acidic Protein (GFAP) in these cells. These results indicate that stabilization of FGF2 by conjugating the IO/HSA NPs can enhance the biological efficacy of FGF2 and its ability to promote hBM-MSC cell proliferation and trilineage differentiation. This new system may benefit future therapeutic use of hBM-MSCs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Divinyl sulfone, contains hydroquinone as inhibitor, ≥96%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous