Structural background for serological cross-reactivity between bacteria of different enterobacterial serotypes.
Electrophoresis (2015-01-30)
Lilla Makszin, Zoltán Péterfi, Ágnes Blaskó, Viktor Sándor, Anikó Kilár, Ágnes Dörnyei, Erzsébet Ősz, Ferenc Kilár, Béla Kocsis
RESUMEN
The structure of the oligosaccharide repeating units of endotoxins from Gram-negative bacteria is characteristic for the different serogroups and serotypes of bacteria. Detailed examination of the cross-reactions of three enterobacterial serotypes, Proteus morganii O34, Escherichia coli O111, and Salmonella enterica sv. Adelaide O35, was performed using sensitive tests (ELISA, immunoblotting). Fine differences between the endotoxins of the bacteria were detected using silver staining of SDS-PAGE gels and chip-technology for the intact lipopolysaccharides (LPSs). The compositions of the O-specific polysaccharides of LPSs extracted from the bacteria were studied, and it was proven that the three cross-reacting bacteria contain O-antigens built from the same monosaccharides, namely colitoses linked to glucose, galactose, and N-acetyl-galactosamine. The NMR and GC-MS studies revealed that the most probable component for the cross-reaction is the rare sugar, colitose.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C