  • Search Results
  • Core component EccB1 of the Mycobacterium tuberculosis type VII secretion system is a periplasmic ATPase.
Core component EccB1 of the Mycobacterium tuberculosis type VII secretion system is a periplasmic ATPase.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-09-24)
Xiao-Li Zhang, De-Feng Li, Joy Fleming, Li-Wei Wang, Ying Zhou, Da-Cheng Wang, Xian-En Zhang, Li-Jun Bi
ABSTRACT
Pathogenic mycobacteria transport virulence factors across their complex cell wall via a type VII secretion system (T7SS)/early secreted antigenic target-6 of kDa secretion system (ESX). ESX conserved component (Ecc) B, a core component of the T7SS architecture, is predicted to be a membrane bound protein, but little is known about its structure and function. Here, we characterize EccB1, showing that it is an ATPase with no sequence or structural homology to other ATPases located in the cell envelope of Mycobacterium tuberculosis H37Rv. We obtained the crystal structure of an EccB1-ΔN72 truncated transmembrane helix and performed modeling and ATP docking studies, showing that EccB1 likely exists as a hexamer. Sequence alignment and ATPase activity determination of EccB1 homologues indicated the presence of 3 conserved motifs in the N- and C-terminals of EccB1-ΔN72 that assemble together between 2 membrane proximal domains of the EccB1-ΔN72 monomer. Models of the EccB1 hexamer show that 2 of the conserved motifs are involved in ATPase activity and form an ATP binding pocket located on the surface of 2 adjacent molecules. Our results suggest that EccB may act as the energy provider in the transport of T7SS virulence factors and may be involved in the formation of a channel across the mycomembrane.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture