• Inicio
  • Resultados de la búsqueda
  • Biomimetic PVPA in vitro model for estimation of the intestinal drug permeability using fasted and fed state simulated intestinal fluids.
Biomimetic PVPA in vitro model for estimation of the intestinal drug permeability using fasted and fed state simulated intestinal fluids.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-04-04)
Elenaz Naderkhani, Terje Vasskog, Gøril Eide Flaten
RESUMEN
A prerequisite for successful oral drug therapy is the drug's ability to cross the gastrointestinal barrier. Considering the increasing number of new chemical entities in modern drug discovery, reliable and fast in vitro models are required for early and efficient prediction of intestinal permeability. To mimic the intestinal environment, use of biorelevant media may provide valuable information on in vivo drug permeation. The present study aims at improving the novel biomimetic phospholipid vesicle-based permeation assay's (PVPAbiomimetic) biorelevance by investigating the applicability of the biorelevant media; fasted state simulated intestinal fluid (FaSSIF) and fed state simulated intestinal fluid (FeSSIF). The FaSSIF and FeSSIF's influence on the permeability of the model drugs acyclovir, indomethacin, griseofulvin and nadolol was then assessed. The barriers' robustness in terms of storage stability was also evaluated. The barriers were found to maintain their integrity in presence of FaSSIF and FeSSIF. The model drugs showed changes in permeability in presence of the different simulated intestinal fluids that were in agreement with previous reports. Moreover, the barrier showed improved storage stability by maintaining its integrity for 6months. Altogether, this study moves the PVPAbiomimetic an important step towards a better in vitro permeability model for use in drug development.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Maleic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)