• Inicio
  • Resultados de la búsqueda
  • CoFe2O4 nano-particles functionalized with 8-hydroxyquinoline for dispersive solid-phase micro-extraction and direct fluorometric monitoring of aluminum in human serum and water samples.
CoFe2O4 nano-particles functionalized with 8-hydroxyquinoline for dispersive solid-phase micro-extraction and direct fluorometric monitoring of aluminum in human serum and water samples.
Analytica chimica acta (2015-06-05)
Hossein Abdolmohammad-Zadeh, Elaheh Rahimpour
RESUMEN
A simple dispersive solid-phase micro-extraction method based on CoFe2O4 nano-particles (NPs) functionalized with 8-hydroxyquinoline (8-HQ) with the aid of sodium dodecyl sulfate (SDS) was developed for separation of Al(III) ions from aqueous solutions. Al(III) ions are separated at pH 7 via complex formation with 8-HQ using the functionalized CoFe2O4 nano-particles sol solution as a dispersed solid-phase extractor. The separated analyte is directly quantified by a spectrofluorometric method at 370nm excitation and 506nm emission wavelengths. A comparison of the fluorescence of Al(III)-8-HQ complex in bulk solution and that of Al(III) ion interacted with 8-HQ/SDS/CoFe2O4 NPs revealed a nearly 5-fold improvement in intensity. The experimental factors influencing the separation and in situ monitoring of the analyte were optimized. Under these conditions, the calibration graph was linear in the range of 0.1-300ngmL(-1) with a correlation coefficient of 0.9986. The limit of detection and limit of quantification were 0.03ngmL(-1) and 0.10ngmL(-1), respectively. The inter-day and intra-day relative standard deviations for six replicate determinations of 150ngmL(-1) Al(III) ion were 2.8% and 1.7%, respectively. The method was successfully applied to direct determine Al(III) ion in various human serum and water samples.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
8-Hydroxyquinoline, ACS reagent, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)