Merck
  • Inicio
  • Resultados de la búsqueda
  • Development and characterization of Haemophilus influenzae type b conjugate vaccine prepared using different polysaccharide chain lengths.
Development and characterization of Haemophilus influenzae type b conjugate vaccine prepared using different polysaccharide chain lengths.
Vaccine (2015-04-25)
R Rana, J Dalal, D Singh, N Kumar, S Hanif, N Joshi, M K Chhikara
RESUMEN
Capsular polysaccharide conjugates of Haemophilus influenzae type b (Hib) are important components of several mono- or multi-valent childhood vaccines. However, their access to the most needy people is limited due to their high cost. As a step towards developing a cost effective and more immunogenic Hib conjugate vaccine, we present a method for the preparation of Hib capsular polysaccharide (PRP)-tetanus toxoid (TT) conjugates using optimized PRP chain length and conjugation conditions. Reactive aldehyde groups were introduced into the polysaccharides by controlled periodate oxidation of the native polysaccharide, which were subsequently covalently linked to hydrazide derivatized tetanus toxoid by means of reductive amination. Native polysaccharides were reduced to average 100 or 50kDa polysaccharide and 10kDa oligosaccharides in a controlled manner. Various conjugates were prepared using Hib polysaccharide and oligosaccharide yielding conjugates with polysaccharide to protein ratios in the range of 0.25-0.5 (w/w) and free saccharide levels of less than 10%. Immunization of Sprague Dawley rats with the conjugates elicited specific antibodies to PRP. The low molecular weight PRP-TT conjugates were found to be more immunogenic as compared to their high molecular weight counterparts and the PRP-TT reference vaccine.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Deuterium oxide, standard, 99.98 atom %±0.01 atom % D
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Deuterium oxide, "100%", 99.990 atom % D
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Deuterium oxide, "100%", ≥99.96 atom % D
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)