Terrein reduces age-related inflammation induced by oxidative stress through Nrf2/ERK1/2/HO-1 signalling in aged HDF cells.
Cell biochemistry and function (2015-09-30)
Young-Hee Lee, Sook-Jeong Lee, Ji-Eun Jung, Jeong-Seok Kim, Nan-Hee Lee, Ho-Keun Yi
RESUMEN
This study investigated whether multiple bioactivity of terrein such as anti-inflammatory and anti-oxidant inhibits age-related inflammation by promoting an antioxidant response in aged human diploid fibroblast (HDF) cells. HDF cells were cultured serially for in vitro replicative senescence. To create the ageing cell phenotype, intermediate stage (PD31) HDF cells were brought to stress-induced premature senescence (SIPS) using hydrogen peroxide (H2 O2). Terrein increased cell viability even with H2O2 stress and reduced inflammatory molecules such as intracellular adhesion molecule-1 (ICAM-1), cyclooxygenase-2 (COX-2), interleukin-1beta (IL-1β) and tumour necrosis factor-alpha (TNF-α). Terrein reduced also phospho-extracellular kinase receptor1/2 (p-EKR1/2) signalling in aged HDF cells. SIPS cells were attenuated for age-related biological markers including reactive oxygen species (ROS), senescence associated beta-galactosidase (SA β-gal.) and the aforementioned inflammatory molecules. Terrein induced the induction of anti-oxidant molecules, copper/zinc-superoxide defence (Cu/ZnSOD), manganese superoxide dismutase (MnSOD) and heme oxygenase-1 (HO-1) in SIPS cells. Terrein also alleviated reactive oxygen species formation through the Nrf2/HO-1/p-ERK1/2 pathway in aged cells. The results indicate that terrein has an alleviative function of age-related inflammation characterized as an anti-oxidant. Terrein might be a useful nutraceutical compound for anti-ageing.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
2′,7′-Dichlorofluorescin diacetate, ≥97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
2-Nitrophenyl β-D-galactopyranoside, ≥98% (enzymatic)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis