• Inicio
  • Resultados de la búsqueda
  • Simultaneous Determination of Aliskiren Hemifumarate, Amlodipine Besylate and Hydrochlorothiazide in Spiked Human Plasma Using UPLC-MS/MS.
Simultaneous Determination of Aliskiren Hemifumarate, Amlodipine Besylate and Hydrochlorothiazide in Spiked Human Plasma Using UPLC-MS/MS.
Journal of chromatographic science (2015-01-13)
Walid M Ebeid, Ehab F Elkady, Asmaa A El-Zaher, Ramzia I El-Bagary, Gabor Patonay
RESUMEN
A sensitive UPLC-MS/MS method was developed and validated for simultaneous estimation of aliskiren hemifumarate (ALS), amlodipine besylate (AML) and hydrochlorothiazide (HCZ) in spiked human plasma using valsartan as an internal standard (IS). Liquid-liquid extraction was used for purification and pre-concentration of analytes. The mobile phase consisted of 0.1% formic acid in ammonium acetate buffer (0.02 M, pH 3.5) and methanol (25:75, v/v), flowing through XBridge BEH (50 × 2.1 mm ID, 5 µm) C18 column, at a flow rate of 0.6 mL min(-1). Multiple reaction monitoring (MRM) transitions were measured using an electrospray source in the positive ion mode for ALS and AML, whereas HCZ and IS were measured in negative ion mode. Validation of the method was performed as per US-FDA guidelines with linearity in the range of 2.0-400.0, 0.3-25.0 and 5.0-400.0 ng mL(-1) for ALS, AML and HCZ, respectively. In human plasma, ALS, AML and HCZ were stable for at least 1 month at -70 ± 5°C and for at least 6 h at ambient temperature. After extraction from plasma, the reconstituted samples of ALS, AML and HCZ were stable in the autosampler at ambient temperature for 6 h. The LC-MS/MS method is suitable for bioequivalence and pharmacokinetic studies of this combination.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C