• Inicio
  • Resultados de la búsqueda
  • Amorphous drug dispersions with mono- and diacyl lecithin: On molecular categorization of their feasibility and UV dissolution imaging.
Amorphous drug dispersions with mono- and diacyl lecithin: On molecular categorization of their feasibility and UV dissolution imaging.
International journal of pharmaceutics (2015-07-02)
Nicolas Gautschi, Peter Van Hoogevest, Martin Kuentz
RESUMEN
There is a growing interest in drug-phospholipid complexes and similar formulations that are mostly solid dispersions with high drug load. This study aims to explore the feasibility of such phospholipid-based solid dispersions as well as to characterize them. A particular aim was to compare monoacyl phosphatidylcholine (PC) with the diacyl excipient. The solid dispersions were manufactured using a solvent evaporation technique and characterized by means of differential scanning calorimetry and X-ray diffractometry. Density functional theory was used to calculate molecular frontier orbitals of the different compounds. Finally, the dissolution properties were analyzed in a flow-through cell by means of UV imaging. It was found that the ability to form solid dispersions with the phospholipids containing amorphous or solubilized drug (at equimolar ratio with the lipid) was dependent on the drug's frontier orbital energy, the enthalpy of fusion, as well as the log P value. In a subsequent dissolution study, UV imaging revealed pronounced surface swelling of the solid dispersions. Only the monoacyl PC was found to substantially enhance in vitro dissolution compared to pure drug. The gained understanding will support a future development of solid drug dispersions using phospholipids as matrix components.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG