Immunoregulatory role of IL-35 in T cells of patients with rheumatoid arthritis.
Rheumatology (Oxford, England) (2015-03-04)
Souichiro Nakano, Shinji Morimoto, Satoshi Suzuki, Hiroshi Tsushima, Kenjiro Yamanaka, Iwao Sekigawa, Yoshinari Takasaki
RESUMEN
IL-35 is the most recently identified member of the IL-12 family. It consists of EBV-induced gene 3 (EBI3) and IL-12α chain p35. We investigated whether IL-35 enhances the in vitro immunosuppressive function of peripheral blood isolated from patients with RA. Peripheral blood was harvested from 17 active and 10 inactive RA patients and IL-35 concentrations were quantified using an ELISA. An expression vector containing IL-35 with a FLAG tag at the carboxyl-terminus was constructed by covalently linking EBI3 and IL-12α (p35). The function of IL-35 was then evaluated in a suppression assay using T cells isolated from human RA patients with CD2, CD3 and CD28 antibodies. Serum IL-35 levels and the number of Treg were decreased significantly in patients with active RA. There was a significant correlation between serum IL-35 and the 28-joint DAS with ESR (DAS28-ESR) in patients with active RA. IL-35 treatment enhanced the regulatory function, suppressing the levels of inflammatory cytokines such as IL-17 and IFN-γ and the cellular growth of effector T cells stimulated by conjugation with CD2, CD3 and CD28. These data revealed that IL-35 might suppress T cell activation during the peripheral immune responses of RA. Therefore our data suggest that IL-35 might have multiple therapeutic targets.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Fluorescein diacetate, used as cell viability stain
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis