Isolation and Characterization of CD271⁺ Stem Cells Derived from Sheep Dermal Skin.
Cells, tissues, organs (2015-05-23)
Roomina Jahroomishirazi, Augustinus Bader, Sabine Ebert, Christian Schmidt, Bita Sedaghati, Michaela Schulz-Siegmund, Matthias Zscharnack
RESUMEN
Mesenchymal stem cells (MSCs) have great promise in the field of regenerative medicine due to their differentiation potential into several lineages. Besides the bone marrow, MSCs can be obtained from the dermis, which represents a large stem cell reservoir in the skin. Sheep provide an appropriate large animal model for preclinical studies. In this study, we focused on the isolation and characterization of MSCs from sheep dermis as an alternative to bone marrow MSCs (bmMSCs). Primary ovine cells were obtained from the dermis for comparison with bone marrow. CD271(+)/45(-) dermal MSCs (CD271-dMSCs), which were sorted by flow cytometry, and plastic-adherent bmMSCs were examined for morphology, proliferation and senescence-associated β-galactosidase activity in both low and high oxygen conditions. CD271 expression on cultured cells was assessed by flow cytometry. Adipogenic and osteogenic potentials of CD271-dMSCs were evaluated by oil red O and von Kossa staining. Chondrogenic capacity of CD271-dMSCs and CD271(+)/CD45(-) bone marrow cells (CD271-bmMSCs) was detected using immunohistochemistry and measurement of sulfated glycosaminoglycans. The cell proliferation assay demonstrated no significant difference between CD271-dMSCs and bmMSCs under low oxygen conditions. Cultured CD271-dMSCs revealed much more CD271 expression compared to CD271-bmMSCs. CD271-dMSCs and CD271-bmMSCs showed basically similar expression of the cartilage-specific proteins aggrecan and collagen type II, although with a stronger staining in CD271-bmMSC-derived cultures. Remarkably, there was co-expression of CD271 and aggrecan during chondrogenic differentiation, suggesting an involvement of CD271 in chondrogenesis. Based on these findings, CD271-dMSCs might serve as an appropriate alternative cell source in preclinical research.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
SAFC
L-Cysteine
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Dexamethasone, meets USP testing specifications