Composite alginate and gelatin based bio-polymeric wafers containing silver sulfadiazine for wound healing.
International journal of biological macromolecules (2015-05-06)
Joshua Boateng, Rocio Burgos-Amador, Obinna Okeke, Harshavardhan Pawar
RESUMEN
Lyophilized wafers comprising sodium alginate (SA) and gelatin (GE) (0/100, 75/25, 50/50, 25/75, 0/100 SA/GE, respectively) with silver sulfadiazine (SSD, 0.1% w/w) have been developed for potential application on infected chronic wounds. Polymer-drug interactions and physical form were characterized by Fourier transform infrared spectroscopy (FTIR) and X-ray diffraction (XRD), respectively, while morphological structure was examined using scanning electron microscopy (SEM). Functional characteristics [(mechanical hardness and adhesion using texture analyzer, and swelling capacity)] of blank wafers were determined in order to select the optimal formulations for drug loading. Finally, the in vitro drug dissolution properties of two selected drug loaded wafers were investigated. There was an increase in hardness and a decrease in mucoadhesion with increasing GE content. FTIR showed hydrogen bonding and electrostatic interaction between carboxyl of SA and amide of GE but no interaction between the polymers and drug was observed, with XRD showing that SSD remained crystalline during gel formulation and freeze-drying. The results suggest that 75/25 SA/GE formulations are the ideal formulations due to their uniformity and optimal mucoadhesivity and hydration. The drug loaded wafers showed controlled release of SSD over a 7h period which is expected to reduce bacterial load within infected wounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Trimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl