PUMA-mediated mitochondrial apoptotic disruption by hypoxic postconditioning.
Apoptosis : an international journal on programmed cell death (2015-06-06)
YuZhen Li, Qi Guo, XiuHua Liu, Chen Wang, DanDan Song
RESUMEN
Postconditioning can reduce ischemia-reperfusion (I/R)-induced cardiomyocyte apoptosis by targeting mitochondria. p53 upregulated modulator of apoptosis (PUMA) is involved in lethal I/R injury. Here, we hypothesized that postconditioning might inhibit mitochondrial pathway-mediated cardiomyocyte apoptosis by controlling PUMA expression. The cultured neonatal rat cardiomyocytes underwent 3 h of hypoxia and 3 h of reoxygenation. Postconditioning consisted of three cycles of 5 min reoxygenation and 5 min hypoxia after prolonged hypoxia. Hypoxic postconditioning reduced the levels of PUMA mRNA and protein. Concomitantly, the loss of mitochondrial membrane potential, cytochrome c release and caspase-3 activation were decreased significantly by postconditioning. Overexpression of PUMA increased greatly not only the number of apoptotic cardiomyocytes, but also the collapse of mitochondrial membrane potential, cytochrome c release and caspase-3 activation under postconditioning condition. The data suggest that reduction of PUMA expression mediates the endogenous cardioprotective mechanisms of postconditioning by disrupting mitochondrial apoptotic pathway.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
SAFC
HEPES
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M