Merck
  • Inicio
  • Resultados de la búsqueda
  • Segregation versus colocalization: orthogonally functionalized binary micropatterned substrates regulate the molecular distribution in focal adhesions.
Segregation versus colocalization: orthogonally functionalized binary micropatterned substrates regulate the molecular distribution in focal adhesions.
Advanced materials (Deerfield Beach, Fla.) (2015-05-20)
Judith Guasch, Bert Conings, Stefanie Neubauer, Florian Rechenmacher, Karen Ende, Claudio G Rolli, Christian Kappel, Viktoria Schaufler, Alexandre Micoulet, Horst Kessler, Hans-Gerd Boyen, Elisabetta Ada Cavalcanti-Adam, Joachim P Spatz
RESUMEN
Orthogonally functionalized binary micropatterned substrates are produced using a novel protocol. The use of adequate peptido-mimetics enables an unprecedented segregation of purified αvβ3 and α5β1 integrins in adjacent microislands and evidences the preference of U2OS cells to colocalize such receptors. Moreover, this tendency can be altered by varying the geometry and composition of the micropatterns.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, BioReagent, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Triton X-100, laboratory grade