Merck
  • Inicio
  • Resultados de la búsqueda
  • Comparative investigations on the biological effects of As (III) and As (V) in clam Ruditapes philippinarum using multiple biomarkers.
Comparative investigations on the biological effects of As (III) and As (V) in clam Ruditapes philippinarum using multiple biomarkers.
Fish & shellfish immunology (2015-09-04)
Chenglong Ji, Hai'e Xu, Qing Wang, Jianmin Zhao, Huifeng Wu
RESUMEN
Inorganic arsenic is a known pollutant with two chemical forms, arsenite (As (III)) and arsenate (As (V)), in marine environment. Clam Ruditapes philippinarum is an important fishery species along the Bohai coast. In this study, the biological effects induced by the two arsenic chemical forms (arsenite and arsenate) were compared using multiple biochemical indices in the digestive glands of clam R. philippinarum. The production of reactive oxygen species, antioxidant enzyme activities and metabolic responses exhibited that both As (III) and As (V) induced immune, oxidative and osmotic stresses in clam digestive glands. The differential metabolic biomarkers, histidine and taurine, indicated the differential responsive mechanisms in osmotic regulation in clam digestive glands. In addition, both arsenic treatments enhanced the anaerobiosis metabolism in clam digestive glands. Overall, this work illustrated that arsenite and arsenate induced similar biological effects in clams, which might be accounted for the biological transformation of arsenate to arsenite in clams.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
2′,7′-Dichlorofluorescein diacetate, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ≥98%
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, puriss. p.a., ACS reagent, ≥98.0% (T)
Sigma-Aldrich
Sodium phosphate tribasic dodecahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Potassium phosphate monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%