  • Search Results
  • Identification of LncRNAs/mRNAs related to endometrium function regulated by Homeobox A10 in Ishikawa cells.
Identification of LncRNAs/mRNAs related to endometrium function regulated by Homeobox A10 in Ishikawa cells.
Cell biology international (2015-03-12)
Ruiyi Lin, Changzhi Zhao, Lu Jing, Shuhong Zhao
ABSTRACT
As a well-known transcription factor, Homeobox A10 (HOXA10) regulates a large number of downstream target genes, leading to the proper function development of endometrium for embryo implantation. The change of HOXA10 gene expression level can alter the expressions of many other genes, including coding and noncoding transcripts. In our study, mRNA and LncRNA expression profiles screening was performed by microarray when the HOXA10 gene expression level increased in Ishikawa cells. A total of 907 mRNAs and 1,026 LncRNAs were identified as differentially expressed transcripts (Fold Change ≥2, P-value <0.05, and Q-value <0.05) between HOXA10 overexpressed and control Ishikawa cells. Further analysis identified that these mRNAs participated in various biological processes, such as blood vessel development, cell adhesion, cell cycle, etc. Also, 14 enhancer-like LncRNAs and 108 LincRNAs with their nearby mRNAs were identified as coregulated transcripts. Our results showed that the mRNA and LncRNA expression profiles differed significantly between the two groups and provided useful information for further studying the molecular mechanisms of HOXA10 in endometrium.
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)