Merck
uPAR Expression Pattern in Patients with Urothelial Carcinoma of the Bladder--Possible Clinical Implications.
PloS one (2015-08-21)
Line Hammer Dohn, Helle Pappot, Benedikte Richter Iversen, Martin Illemann, Gunilla Høyer-Hansen, Ib Jarle Christensen, Peter Thind, Lisbeth Salling, Hans von der Maase, Ole Didrik Laerum
RESUMEN
The objective of the present study was to confirm the expression and localisation pattern of the urokinase-type plasminogen activator receptor (uPAR) focusing on its possible clinical relevance in patients with urothelial neoplasia of the bladder. uPAR is a central molecule in tissue remodelling during cancer invasion and metastasis and is an established prognostic marker in various cancer diseases other than bladder cancer. Formalin-fixed and paraffin-embedded tumour-tissue blocks from 186 patients treated with radical cystectomy were analysed. uPAR expression was scored as either negative or positive as well as by the actual score. Separate scores were obtained for cancer cells, macrophages and myofibroblasts at the invasive front and in tumour core. We were able to confirm, in an independent patient cohort, the tissue expression and localisation pattern of uPAR as investigated by Immunohistochemistry as well as a significant association between uPAR positivity and increasing tumour stage and tumour grade. This demonstrates the robustness of our previous and current findings. In addition the association between uPAR positive myofibroblasts and poor survival was reproduced. The highest hazard ratios for survival were seen for uPAR positive myofibroblasts both at the invasive front and in tumour core. Evaluating uPAR expression by the actual score showed a significant association between uPAR positive myofibroblasts in tumour core and an increased risk of cancer specific mortality. Our investigations have generated new and valuable biological information about the cell types being involved in tumour invasion and progression through the plasminogen activation system.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%