Quercetin and Resveratrol Decrease the Inflammatory and Oxidative Responses in Human Ocular Surface Epithelial Cells.
Investigative ophthalmology & visual science (2015-06-13)
Antonio Abengózar-Vela, Margarita Calonge, Michael E Stern, María Jesús González-García, Amalia Enríquez-De-Salamanca
RESUMEN
To determine the anti-inflammatory and antioxidant effects of quercetin (QCT) and/or resveratrol (RES) on human conjunctival (IOBA-NHC) and corneal (HCE) epithelial cell lines. IOBA-NHC and HCE cells were treated with QCT (0.5-25 μM), RES (0.5-50 μM) and a low-dose mixture of QCT (0.5 μM) and RES (5 μM) (QCT+RES) and stimulated with either TNF-α or ultraviolet (UV)-B radiation. Cytokine production (IL-6, IL-8, IP-10, and VEGF) was analyzed by an immune bead-based array, and intracellular reactive oxygen species (ROS) production was determined by a H2DCF-DA dye assay. Stimulation of IOBA-NHC and HCE cells with TNF-α induced an increase of IL-6, IL-8, and IP-10 secretion in both cell lines. Quercetin and RES decreased IL-6 and IP-10 secretion in a dose-dependent manner in both cell lines. Interleukin-8 secretion was also inhibited in a dose-dependent manner by QCT in HCE, but only at 20 and 25 μM QCT and 50 μM RES in IOBA-NHC and at 50 μM RES in HCE. QCT+RES decreased IL-6 and IL-8 secretion (P < 0.01 and P < 0.05, respectively) in IOBA-NHC cells. Ultraviolet-B induced a significant increase of ROS in both cell lines (P < 0.01 and P < 0.001 for IOBA-NHC and HCE cells, respectively), which was significantly decreased in a dose-dependent manner by QCT and RES in HCE cells. Reactive oxygen species production in IOBA-NHC cells was inhibited (P < 0.05) by 50 μM RES. Quercetin and RES have anti-inflammatory and antioxidant effects on IOBA-NHC and HCE cells. These in vitro data suggest that both polyphenols may have a therapeutic potential in the treatment of inflammatory ocular surface diseases.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
SAFC
HEPES
SAFC
L-Glutamine
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture