Microfluidic sampling system for tissue analytics.
Biomicrofluidics (2015-10-01)
A Hokkanen, I Stuns, P Schmid, A Kokkonen, F Gao, A Steinecker, J Budczies, P Heimala, L Hakalahti
RESUMEN
We have developed a microfluidics based sampling system for tissue analytics. The proof-of-concept of the sampling system was demonstrated by extracting lipid samples from tissue biopsies. The sample collection system consists of a disposable silicon based multiport microneedle integrated with polymer microfluidics. The polymethyl methacrylate polymer microfluidic chip has a 10 μl sample reservoir and actuation membranes for liquid pumping. A special automated robotic system was developed to control the positioning of the needle and the sampling procedure on preselected spots on the tissue. Real breast cancer tissue samples were used to test the feasibility of the sampling system. We successfully measured indicative cancer biomarkers from the tissue surface. Phosphatidylcholine and phosphoethanolamine were extracted from the tissue membrane with methyl tert-butyl ether solvent and detected by mass spectrometry. In the future, this tool could be used in characterization of preoperative biopsies and tumour tissues removed during surgery.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%