Merck
  • Inicio
  • Resultados de la búsqueda
  • Preliminary study on brominated dioxins/furans and hydroxylated/methoxylated PBDEs in Baltic cod (Gadus morhua) liver. Comparison to the levels of analogue chlorinated co-occurring pollutants.
Preliminary study on brominated dioxins/furans and hydroxylated/methoxylated PBDEs in Baltic cod (Gadus morhua) liver. Comparison to the levels of analogue chlorinated co-occurring pollutants.
Marine pollution bulletin (2015-05-24)
Marek Roszko, Krystyna Szymczyk, Małgorzata Rzepkowska, Renata Jędrzejczak
RESUMEN
Low pg levels of PBDD/Fs in 0.8-38.9 pg g(-1) (fat) range (mean 14.2 pg g(-1)) observed in the tested cod liver samples were significantly lower than levels of their chlorinated analogues (25.1-95.9 range, 37.7 mean pg g(-1) fat). Brominated furans dominated the congener profile. Elevated levels of OH-/MeO-BDE were however observed with mean concentrations at 2769±1319 and 5441±2918 pg g(-1) (fat) respectively. The observed congener profiles with the predominance of 6-OH-BDE 47 and the relatively high proportion between the native and hydroxylated compounds indicate on natural origin of those compounds or at least presence of various exposure sources. A large fraction of studied samples did not comply with the currently standing EU regulations concerning PCBs and chlorinated dioxins (11%), especially as regards non-dioxin like PCBs (88%).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Carbon, mesoporous, average pore diameter 100±10 Å (typical), >99.95% trace metals basis
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methane, electronic grade, ≥99.998%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Carbon, mesoporous, nanopowder, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%