Antioxidant Capacities and Analysis of Phenolic Compounds in Three Endemic Nolana Species by HPLC-PDA-ESI-MS.
Molecules (Basel, Switzerland) (2015-06-26)
Mario J Simirgiotis, Julio Benites, Carlos Areche, Beatriz Sepúlveda
RESUMEN
The antioxidant features, polyphenolic composition and chromatographic fingerprints of the aerial parts from three Chilean endemic plants from the Paposo Valley located on the cost of the Atacama Desert were investigated for the first time using high pressure liquid chromatography coupled with photodiode array detector and electrospray ionization mass analysis (HPLC-PDA-ESI-MS) and spectroscopic methods. The phenolic fingerprints obtained for the plants were compared and correlated with the antioxidant capacities measured by the bleaching of the DPPH radical, the ferric reducing antioxidant power (FRAP) and quantification of the total content of phenolics and flavonoids measured by spectroscopic methods. Thirty phenolics were identified for the first time for these species, mostly phenolic acids, flavanones, flavonols and some of their glycoside derivatives, together with three saturated fatty acids (stearic, palmitic and arachidic acids). Nolana ramosissima showed the highest antioxidant activity (26.35 ± 1.02 μg/mL, 116.07 ± 3.42 μM Trolox equivalents/g dry weight and 81.23% ± 3.77% of inhibition in the DPPH, FRAP and scavenging activity (SA) assays, respectively), followed by  N. aplocaryoides (85.19 ± 1.64 μg/mL, 65.87 ± 2.33 μM TE/g DW and 53.27% ± 3.07%) and N. leptophylla (124.71 ± 3.01, 44.23 ± 5.18 μM TE/g DW and 38.63% ± 1.85%).
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Luperox® TBH70X, tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Xanthine, ≥99%