• Inicio
  • Resultados de la búsqueda
  • Facultative Anaerobe Caldibacillus debilis GB1: Characterization and Use in a Designed Aerotolerant, Cellulose-Degrading Coculture with Clostridium thermocellum.
Facultative Anaerobe Caldibacillus debilis GB1: Characterization and Use in a Designed Aerotolerant, Cellulose-Degrading Coculture with Clostridium thermocellum.
Applied and environmental microbiology (2015-06-07)
Scott Wushke, David B Levin, Nazim Cicek, Richard Sparling
RESUMEN
Development of a designed coculture that can achieve aerotolerant ethanogenic biofuel production from cellulose can reduce the costs of maintaining anaerobic conditions during industrial consolidated bioprocessing (CBP). To this end, a strain of Caldibacillus debilis isolated from an air-tolerant cellulolytic consortium which included a Clostridium thermocellum strain was characterized and compared with the C. debilis type strain. Characterization of isolate C. debilis GB1 and comparisons with the type strain of C. debilis revealed significant physiological differences, including (i) the absence of anaerobic metabolism in the type strain and (ii) different end product synthesis profiles under the experimental conditions used. The designed cocultures displayed unique responses to oxidative conditions, including an increase in lactate production. We show here that when the two species were cultured together, the noncellulolytic facultative anaerobe C. debilis GB1 provided respiratory protection for C. thermocellum, allowing the synergistic utilization of cellulose even under an aerobic atmosphere.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98%
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Sorbitol, 99%
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(−)-Ribose, ≥99%
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-Sorbitol, for molecular biology, ≥98%
Sigma-Aldrich
L-Rhamnose, natural sourced, 99%, FG
Sigma-Aldrich
D-(+)-Mannose, synthetic, ≥99%
Sigma-Aldrich
D-Sorbitol, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-(−)-Arabinose, ≥98%
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
D-(+)-Mannose, wood, ≥99%
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, BioXtra, ≥98%
Sigma-Aldrich
D-Sorbitol, FCC, FG
Sigma-Aldrich
D-(−)-Ribose, suitable for cell culture, BioReagent