• Inicio
  • Resultados de la búsqueda
  • Inhibitory effects of high stability fucoxanthin on palmitic acid-induced lipid accumulation in human adipose-derived stem cells through modulation of long non-coding RNA.
Inhibitory effects of high stability fucoxanthin on palmitic acid-induced lipid accumulation in human adipose-derived stem cells through modulation of long non-coding RNA.
Food & function (2015-06-10)
Wen-Chuan Lin, Ping-Hsiao Shih, Weu Wang, Chi-Hao Wu, Shih-Min Hsia, Hsian-Jenn Wang, Pai-An Hwang, Chuan-Yu Wang, Shu-Huey Chen, Yung-Ting Kuo
RESUMEN
Obesity is a serious worldwide disease, which is growing in epidemic proportions. Adipose-derived stem cells (ADSCs) are characterized as a source of mesenchymal stem cells that have acted as a potential application for regeneration. Recently, seaweeds rich in flavonoids and polysaccharides have been supposed to show the ability to modulate risk factors for obesity and related diseases. In the present study, we investigated the anti-obesity properties of high stability fucoxanthin (HS-Fx) derived from brown seaweeds on the adipogenesis of ADSCs upon treatment with palmitic acid (PA). First, we showed the differentiation capability of ADSCs from morbid obesity patients to transform into different cell types. Second, we found that the co-treatment of ADSCs with HS-Fx and PA showed no significant cytotoxicity against ADSCs, but PA induced the elevation of reactive oxygen species (ROS) and lipid droplet accumulation was abolished. Thirdly, the PA-mediated down-regulation of lipid metabolism genes was reversed by the treatment of HS-Fx. By long non-coding RNAs (lncRNAs) screening, we found that PA-induced increases in the targeted lncRNAs were also decreased upon treatment with HS-Fx. On Silencing, these lncRNAs corresponded to the decrease in the lipid droplet accumulation of ADSCs induced by PA. ADSCs from obese patients would be direct and meaningful model cells to investigate the development of obesity-related diseases and their treatments, rather than cell lines from other species. HS-Fx showed anti-obesity capability through modulating the elevation of ROS, down-regulation of lipid metabolism genes induced by PA, and upstream signaling, which might be critically resulted from the expression of lncRNAs.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Palmitic acid, ≥95%, FCC, FG
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)