  • Search Results
  • Antimicrobial susceptibility of Canadian isolates of Helicobacter pylori in Northeastern Ontario.
Antimicrobial susceptibility of Canadian isolates of Helicobacter pylori in Northeastern Ontario.
The Canadian journal of infectious diseases & medical microbiology = Journal canadien des maladies infectieuses et de la microbiologie medicale (2015-08-04)
Nelson F Eng, Gustavo Ybazeta, Katrina Chapman, Nya L Fraleigh, Rebecca Letto, Eleonora Altman, Francisco Diaz-Mitoma
ABSTRACT
Helicobacter pylori plays a significant role in gastritis and ulcers. It is a carcinogen as defined by the WHO, and infection can result in adenocarcinomas and mucosa-associated lymphoid tissue lymphomas. In Canada, rates of antimicrobial resistance are relatively unknown, with very few studies conducted in the past 15 years. To examine rates of resistance in Sudbury, Ontario, compare antimicrobial susceptibility methods and attempt to determine the molecular basis of antibiotic resistance. Patients attending scheduled visits at Health Sciences North (Sudbury, Ontario) provided gastric biopsy samples on a volunteer basis. In total, 20 H pylori isolates were collected, and antimicrobial susceptibility testing (on amoxicillin, tetracycline, metronidazole, ciprofloxacin, levofloxacin and clarithromycin) was conducted using disk diffusion and E-test methods. Subsequently, genomic DNA from these isolates was sequenced to detect mutations associated with antimicrobial resistance. Sixty-five percent of the isolates were found to be resistant to at least one of the listed antibiotics according to E-test. Three isolates were found to be resistant to ≥3 of the above-mentioned antibiotics. Notably, 25% of the isolates were found to be resistant to both metronidazole and clarithromycin, two antibiotics that are normally prescribed as part of first-line regimens in the treatment of H pylori infections in Canada and most of the world. Among the resistant strains, the sequences of 23S ribosomal RNA and gyrA, which are linked to clarithromycin and ciprofloxacin/levofloxacin resistance, respectively, revealed the presence of known point mutations associated with antimicrobial resistance. In general, resistance to metronidazole, ciprofloxacin/levofloxacin and clarithromycin has increased since the studies in the early 2000s. These results suggest that surveillance programs of H pylori antibiotic resistance may need to be revisited or improved to prevent antimicrobial therapy failure.
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
3-Methyl-1-butanol, anhydrous, ≥99%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
3-Methylbutanol, BioReagent, for molecular biology, ≥98.5%
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, ≥99% (T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)