Merck
ICA1L forms BAR-domain complexes with PICK1 and is crucial for acrosome formation in spermiogenesis.
Journal of cell science (2015-08-27)
Jing He, Mengying Xia, Wai Hung Tsang, King Lau Chow, Jun Xia
RESUMEN
Mutations in the Pick1 gene cause globozoospermia, a male infertility disorder, in both mice and humans. PICK1 is crucial for vesicle trafficking, and its deficiency in sperm cells leads to abnormal vesicle trafficking from the Golgi to the acrosome. This eventually disrupts acrosome formation and leads to male infertility. Here, we identified ICA1L, which has sequence similarities to ICA69 (also known as ICA1), as a new BAR-domain binding partner of PICK1. ICA1L is expressed in testes and brain, and is the major binding partner for PICK1 in testes. ICA1L and PICK1 are highly expressed in spermatids and trafficked together at different stages of spermiogenesis. ICA1L-knockout mice were generated by CRISPR-Cas technology. PICK1 expression was reduced by 80% in the testes of male mice lacking ICA1L. Sperm from ICA1L-knockout mice had abnormalities in the acrosome, nucleus and mitochondrial sheath formation. Both total and mobile sperm numbers were reduced, and about half of the remaining sperm had the characteristics of globozoospermia. These defects ultimately resulted in reduced fertility of male ICA1L-knockout mice, and ICA69/ICA1L-double knockout male mice were sterile.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Sucrose, analytical standard