Resolving the daratumumab interference with blood compatibility testing.
Transfusion (2015-03-13)
Claudia I Chapuy, Rachel T Nicholson, Maria D Aguad, Bjoern Chapuy, Jacob P Laubach, Paul G Richardson, Parul Doshi, Richard M Kaufman
RESUMEN
Daratumumab (DARA), a promising novel therapy for multiple myeloma, is an IgG1κ monoclonal antibody that recognizes CD38 on myeloma cells. During routine compatibility testing, we observed that the plasma of five of five DARA-treated patients demonstrated a positive antibody screen and panreactivity on red blood cell (RBC) panel testing. We hypothesized that the observed panreactivity reflected DARA binding to CD38 on reagent RBCs, and we investigated methods to prevent this binding. DARA binding to CD38+ or CD38- HL60 cells was assessed by flow cytometry. To remove cell surface CD38, cells were incubated with dithiothreitol (DTT) or trypsin. Soluble CD38 or anti-DARA was used to neutralize DARA in solution. Routine blood bank serologic methods were used to test samples from DARA-treated patients and normal plasma samples spiked with DARA and/or alloantibodies. Normal plasma samples spiked with DARA (0.1-10 µg/mL) and incubated with reagent RBCs recapitulated the interference observed with samples from DARA-treated patients. Flow cytometry experiments confirmed DARA binding to CD38+ HL60 cells, but not to CD38- controls. DTT treatment of CD38+ HL60 cells reduced DARA binding by 92% by denaturing cell surface CD38. Treating DARA-containing plasma with soluble CD38 or anti-DARA idiotype also inhibited DARA binding. DARA causes panreactivity in vitro by binding to CD38 on reagent RBCs. Treating reagent RBCs with DTT is a robust method to negate the DARA interference, enabling the safe provision of blood to DARA-treated patients. Because DTT denatures Kell antigens, K- units are provided to these patients.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
HEPES
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
HEPES
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
L-Glutamine