Merck
  • Inicio
  • Resultados de la búsqueda
  • Liquid Chromatography with Amperometric Detection at a Silver Based Detector for the Determination of Thiocompounds: Application to the Assay of Thiopurine Antimetabolites in Urine.
Liquid Chromatography with Amperometric Detection at a Silver Based Detector for the Determination of Thiocompounds: Application to the Assay of Thiopurine Antimetabolites in Urine.
Analytical chemistry (2015-05-30)
Nurgul Karadas-Bakirhan, Ahmad Sarakbi, Marie Vandeput, Sibel A Ozkan, Jean-Michel Kauffmann
RESUMEN
A silver amperometric detector coupled to liquid chromatography (LC) was used for the determination of 6-thioguanine (6-TG) and two of its metabolites, thiouric acid (TU) and 2-amino-6-mercaptopurine riboside (6-TGR). The silver detector coupled to LC operated at a low applied potential (0.08 V vs Ag/AgCl) and offered a chromatogram with peak responses corresponding to molecules interacting with silver, namely, chloride ions and small soluble biothiols in addition to the organothiol drug compounds investigated. Online electrochemical surface cleaning permitted the improvement of the repeatability and peak shape of the recorded signal compared to direct current amperometric detection (AD) when operating in chloride containing media. The studied molecules were eluted isocratically within 5 min on a reversed-phase C18 column without interference from endogenous biothiols present in urine samples. Diluted urine samples (1:1) were directly injected in the LC setup; a linear calibration curve was obtained between peak area and analyte concentration between 0.1 and 10 μM for all the studied molecules. Limits of detection (LODs) were 0.03, 0.008, and 0.01 μM, and the limits of quantification (LOQs) were 0.1, 0.02, and 0.03 μM for TU, 6-TG, and 6-TGR, respectively. Within-day RSDs were 2%, 0.8%, and 1% and between-day RSDs were 2%, 0.9%, 2% for TU, 6-TG, and 6-TGR, respectively. Recoveries in spiked urine were 99.8%, 99.9%, and 99.0% for TU, 6-TG, and 6-TGR, respectively.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
DL-Homocysteine, ≥95% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
2-Amino-6-methylmercaptopurine, ≥95%
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture