• Inicio
  • Resultados de la búsqueda
  • A quantitative LC-MS/MS method for determining ipragliflozin, a sodium-glucose co-transporter 2 (SGLT-2) inhibitor, and its application to a pharmacokinetic study in rats.
A quantitative LC-MS/MS method for determining ipragliflozin, a sodium-glucose co-transporter 2 (SGLT-2) inhibitor, and its application to a pharmacokinetic study in rats.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-07-26)
Shinji Kobuchi, Yukako Ito, Kyoka Yano, Toshiyuki Sakaeda
RESUMEN
Ipragliflozin is a highly potent and selective sodium-dependent glucose co-transporter-2 (SGLT2) inhibitor, a novel class of hypoglycemic agents. The aim of the present study was to establish a new highly sensitive and rapid liquid chromatography-tandem mass spectrometry (LC-MS/MS) method for the quantitative analysis of ipragliflozin in rat plasma and apply this method to a pharmacokinetic study in rats. Empagliflozin was used as an internal standard (I.S.) and liquid-liquid extraction was conducted using tert-butyl methyl ether. Chromatographic separation was accomplished on a Quicksorb ODS (2.1mm i.d.×150mm, 5μm in size) with acetonitrile/0.1% formic acid (90:10, v/v) at a flow rate of 0.2mL/min. An API 3200 triple quadrupole mass spectrometer operating in the positive electrospray ionization mode with multiple reaction monitoring was used to detect ipragliflozin and I.S. transitions: m/z 422.0 [M+NH4](+)→151.0 for ipragliflozin and m/z 451.2 [M+H](+)→71.0 for I.S. Inter- and intra-day accuracies and precisions were within ±15%. This validated method was successfully applied to a pharmacokinetic study of ipragliflozin in rats. This assay method may contribute to assessment of novel SGLT2 inhibitors using the rat as an animal model.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Acetonitrile solution, contains 0.1 % (v/v) ammonium hydroxide
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C