• Inicio
  • Resultados de la búsqueda
  • A flow cytometer-based method to simultaneously assess activity and selectivity of compounds against the intracellular forms of Trypanosoma cruzi.
A flow cytometer-based method to simultaneously assess activity and selectivity of compounds against the intracellular forms of Trypanosoma cruzi.
Acta tropica (2015-08-15)
Cristian Gabriel Miranda, Maria Elisa Solana, Maria de Los Angeles Curto, Estela Maria Lammel, Alejandro Gabriel Schijman, Catalina Dirney Alba Soto
RESUMEN
Chagas disease is a major unsolved health issue in Latin America and an emerging threat worldwide. New drugs are urgently needed for chemotherapy as those available (benznidazole and nifurtimox) have variable efficacy and elevated toxicity. Efforts are actually oriented to improve tools and technologies (e.g. transgenic parasites, flow cytometry or image-based systems) for the screening of large numbers of candidate compounds for their activity against Trypanosoma cruzi (T. cruzi). Methods that test drug efficacy and selectivity in the same assay are suitable to accelerate the process of drug discovery. Here, we developed a GFP expressing T. cruzi from a moderate virulence stock and confirmed that the transgenic parasite retained the biological characteristics of the parental strain. With this tool, we established a flow cytometer-based method to simultaneously test drug activity against intracellular amastigotes and toxicity to the host cell. This one-step procedure allows determining the selectivity index of the tested compound in a sensitive and accurate manner even with low infection rates. This method can provide additional information on the interactions between drug, parasites and host cell and could be adapted to other trypanosomatids and protozoa with intracellular multiplication.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder