• Inicio
  • Resultados de la búsqueda
  • Identification of Missing Proteins Defined by Chromosome-Centric Proteome Project in the Cytoplasmic Detergent-Insoluble Proteins.
Identification of Missing Proteins Defined by Chromosome-Centric Proteome Project in the Cytoplasmic Detergent-Insoluble Proteins.
Journal of proteome research (2015-06-26)
Yang Chen, Yaxing Li, Jiayong Zhong, Jing Zhang, Zhipeng Chen, Lijuan Yang, Xin Cao, Qing-Yu He, Gong Zhang, Tong Wang
RESUMEN
Finding protein evidence (PE) for protein coding genes is a primary task of the Phase I Chromosome-Centric Human Proteome Project (C-HPP). Currently, there are 2948 PE level 2-4 coding genes per neXtProt, which are deemed missing proteins in the human proteome. As most samples prepared and analyzed in the C-HPP framework were focusing on detergent soluble proteins, we posit that as a natural composition the cytoplasmic detergent-insoluble proteins (DIPs) represent a source of finding missing proteins. We optimized a workflow and separated cytoplasmic DIPs from three human lung and three human hepatoma cell lines via differential speed centrifugation. We verified that the detergent-soluble proteins (DSPs) could be sufficiently depleted and the cytoplasmic DIP isolation was partially reproducible with Spearman r > 0.70 according to two independent SILAC MS experiments. Through label-free MS, we identified 4524 and 4156 DIPs from lung and liver cells, respectively. Among them, a total of 23 missing proteins (22 PE2 and 1 PE4) were identified by MS, and 18 of them had translation evidence; in addition, six PE5 proteins were identified by MS, three with translation evidence. We showed that cytoplasmic DIPs were not an enrichment of transmembrane proteins and were chromosome-, cell type-, and tissue-specific. Furthermore, we demonstrated that DIPs were distinct from DSPs in terms of structural and physical-chemical features. In conclusion, we have found 23 missing proteins and 6 PE5 proteins from the cytoplasmic insoluble proteome that is biologically and physical-chemically different from the soluble proteome, suggesting that cytoplasmic DIPs carry comprehensive and valuable information for finding PE of missing proteins. The mass spectrometry proteomics data have been deposited to the ProteomeXchange Consortium with the data set identifier PXD001694.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl