Insight into the retention mechanism on immobilized artificial membrane chromatography using two stationary phases.
Journal of chromatography. A (2015-04-26)
Fotios Tsopelas, Niki Malaki, Theodosia Vallianatou, Marios Chrysanthakopoulos, Demetris Vrakas, Maria Ochsenkühn-Petropoulou, Anna Tsantili-Kakoulidou
RESUMEN
The retention behavior of sixty structurally diverse drugs on two immobilized artificial membrane (IAM) columns, IAM.PC.MG and IAM.PC.DD2 types, at two pH values, 7.4 and 5.5, was established. Extrapolated to pure aqueous phase retention factors, logkw(IAM), were determined and the role of acetonitrile as organic modifier was explored, considering the relationships with the slopes, S, of the extrapolation procedure. Good interrelations between retention factors on the two IAM stationary phases were observed, although logkw(IAM.PC.DD2) values are generally higher than logkw(IAM.PC.MG). In order to investigate the underlying retention mechanism, relationships between IAM retention factors and lipophilicity, expressed as logP or logD at pH 7.4 were established. Electrostatic interactions were considered by introducing the positively and negatively charged molecular fractions as additional parameters in the logkw(IAM)/logD relationships. The positive contribution of these fractions supported the involvement of the electrostatic interactions in the retention mechanism. Special attention was given to the retention behavior of zwitterionic compounds and for compounds with special structural characteristics.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
SAFC
L-Cystine
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
SAFC
L-Cystine
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis