• Inicio
  • Resultados de la búsqueda
  • Survey of Omp19 immunogenicity against Brucella abortus and Brucella melitensis: influence of nanoparticulation versus traditional immunization.
Survey of Omp19 immunogenicity against Brucella abortus and Brucella melitensis: influence of nanoparticulation versus traditional immunization.
Veterinary research communications (2015-09-24)
Morteza Abkar, Abbas Sahebghadam Lotfi, Jafar Amani, Khadijeh Eskandari, Mehdi Fasihi Ramandi, Jafar Salimian, Gholamreza Nikbakht Brujeni, Saeed Alamian, Mehdi Kamali, Hamid Koushki
RESUMEN
Brucellosis is the most common zoonotic bacterial disease. Prevention of human brucellosis is achieved through pasteurization of dairy products, appropriate sanitation and vaccination of domestic animals against the Brucella species. B. abortus unlipidated 19 kDa outer membrane protein (U-Omp19) is a promising candidate for a subunit vaccine against brucellosis. This study investigates immunogenicity of Omp19 alone and with Freund's adjuvant (Omp19-IFA) and N-trimethyl chitosan (TMC/Omp19) nanoparticles, as well as the effect of Omp19 administration route on immunological responses and protection. The omp19 gene was expressed in E. coli BL21 (DE3). After purification, the recombinant Omp19 was loaded onto TMC nanoparticles by ionic gelation with tripolyphosphate. Particle size and loading efficiency of the nanoparticles were determined. Omp19-IFA was administered intraperitoneally while TMC/Omp19 nanoparticles were administered orally and intraperitoneally. The results indicated that intraperitoneal (i.p.) immunization by Omp19-IFA and TMC/Omp19 nanoparticles induced Th1 and Th2 immune responses, respectively, whereas oral immunization of TMC/Omp19 nanoparticles induced a mixed Th1/Th17 immune response. Moreover, oral immunization increased IgA levels in feces. Immunized mice were challenged with virulent B. melitensis 16 M and B. abortus 544. Oral immunization with TMC/Omp19 nanoparticles induced a remarkably high protection level against B. melitensis and B. abortus. The results showed that immunization route has a pivotal role in immune response polarization and protective efficiency of Omp19 antigen. Also, it was deduced that the higher protection level achieved through oral administration of TMC/Omp19 nanoparticles may be due to the elicited Th17 response.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
SAFC
HEPES
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium tripolyphosphate, technical grade, 85%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
Sodium triphosphate pentabasic, purum p.a., ≥98.0% (T)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
SAFC
HEPES
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%