Evaluating the translocation properties of a new nuclear targeted penetrating peptide using two fluorescent markers.
Journal of drug targeting (2015-01-23)
Rui Zhang, Xu-Zhong Yang, Jia-Wei Wang, Long-Yun Fu, Jian Zhao, Fu-Jun Wang
RESUMEN
Human-derived cell penetrating peptides (CPPs) have attracted much more attentions than other CPPs which are limited by their potential toxicity and immunogenicity. Previously, we identified a novel human-originated CPP (named heparin-binding domain (HBD) in this article), which derived from the C-terminus of human extracellular superoxide dismutase, and demonstrated HBD is an efficient vector for delivering exogenous drug molecules such as apoptin into HeLa cells. In this study, we found this novel CPP showed differentiated efficiency in several tested cell lines. Heparin competitive inhibition experiment and heparanase pre-incubation experiment showed cell surface polysaccharides play an important role for the transmembrane transport. The results of endocytosis inhibitors suggested that HBD penetrates the cell membrane via a direct translocation, which is different from that of TAT, a classical clathrin-mediated endocytosis. HBD could deliver up to 90 kD protein cargoes into cells. Different conjugated modes with cargo molecules greatly affect their translocation efficiency. HBD also showed significant nuclear transport capacity when it was incubated with HeLa cells. Furthermore, the core region for HBD possessing membrane-penetrating ability was identified by deletion analyses. These results would be helpful for developing HBD as a new nuclear delivery tool for therapeutic biomolecules.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl