Evaluation of etanercept degradation under oxidative stress and potential protective effects of various amino acids.
International journal of pharmaceutics (2015-07-19)
Jun Yeul Lim, Nam Ah Kim, Dae Gon Lim, Ki Hyun Kim, Shavron Hada, Seong Hoon Jeong
RESUMEN
To evaluate the oxidative stability of proteins, a model protein, etanercept, was exposed to oxidative stress conditions using hydrogen peroxide. Various amino acids were also evaluated on their antioxidant effect. Transition temperature (Tm), secondary structural content, hydrodynamic size, and aggregation and fragmentation of etanercept in solution were assessed using dynamic light scattering (DLS), size exclusion chromatography (SEC), differential scanning calorimetry (DSC), and far-UV circular dichroism (CD). Sample solutions were stored at 4 °C, 20 °C, and 40 °C under oxidative stress. The DLS results exhibited a decrease in the Z-average and intensity peak size of etanercept during the storage, suggesting fragmentation issues rather than aggregation by oxidation. The SEC results exhibited an increase in fragmentation and a decrease in aggregation and monomer content. The monomer content remained higher in histidine than in other amino acids, followed by methionine. There were three Tm of etanercept that were selected as key parameters of conformational stability. Oxidized samples exhibited a significant decrease in Tm values, indicating decreased conformational stability. Methionine exhibited the highest values in Tm1, followed by histidine. The CD spectrum exhibited one unique negative peak of etanercept without amino acids, and changed with oxidation. Only methionine exhibited an enhancement of the stability. All four biophysical analyses results suggest that the histidine and methionine provide a protective effect in the protein solution against oxidative stress. However, histidine was effective as an antioxidant but methionine showed highly enhanced conformational and secondary structural stability.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
L-Arginine monohydrochloride, reagent grade, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
SAFC
L-Methionine
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SAFC
L-Histidine