Novel antisense therapeutics delivery systems: In vitro and in vivo studies of liposomes targeted with anti-CD20 antibody.
Journal of controlled release : official journal of the Controlled Release Society (2015-11-21)
Justyna M Meissner, Monika Toporkiewicz, Aleksander Czogalla, Lucyna Matusewicz, Kazimierz Kuliczkowski, Aleksander F Sikorski
RESUMEN
Antisense gene therapy using molecules such as antisense oligodeoxynucleotides, siRNA or miRNA is a very promising strategy for the treatment of neoplastic diseases. It can be combined with other treatment strategies to enhance therapeutic effect. In acute leukemias, overexpression of the antiapoptotic gene BCL2 is observed in more than 70% of cases. Therefore, reduction of the Bcl-2 protein level could, in itself, prevent the development of cancer or could possibly help sensitize cancer cells to apoptosis inducers. The main objective of our work is to develop therapeutic liposome formulations characterized by high transfection efficiency, stability in the presence of serum, as well as specificity and toxicity for target (leukemic) cells. Each of our liposomal formulations consists of a core composed of antisense oligonucleotides complexed by either cationic lipid, DOTAP, or a synthetic polycation, polyethyleneimine, encapsulated within liposomes modified with polyethylenoglycol. In addition, the liposomal shells are enriched with covalently-bound antibodies recognizing a well characterized bio-marker, CD20, exposed on the surface of leukemia cells. The resulting immunoliposomes selectively and effectively reduced the expression of BCL2 in target cells. Model animal experiments carried out on mice-engrafted tumors expressing the specific marker showed high efficiency of the liposome formulations against specific tumor development. In conclusion, we show that lipid formulations based on a polyplex or lipoplex backbone additionally equipped with antibodies are promising non-viral vectors for specific oligonucleotide transfer into human tumor cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%